missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-phenoxypropane, 98%

Catalog No. AAL1249414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1249414 25 g
AAL1249422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1249414 Supplier Thermo Scientific Chemicals Supplier No. L1249414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 588-63-6 | C9H11BrO | 215.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 588-63-6
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
MDL Number MFCD00000256
InChI Key NIDWUZTTXGJFNN-UHFFFAOYSA-N
Synonym 3-bromopropoxy benzene, 3-phenoxypropyl bromide, benzene, 3-bromopropoxy, 3-bromopropyl phenyl ether, 1-bromo-3-phenoxypropane, ether, 3-bromopropyl phenyl, gamma-phenoxypropyl bromide, phenoxypropyl bromide, 3-bromo-1-phenoxypropane, .gamma.-phenoxypropyl bromide
PubChem CID 68522
IUPAC Name 3-bromopropoxybenzene
SMILES BrCCCOC1=CC=CC=C1

Specifications

Melting Point 10°C to 11°C
Density 1.365
Boiling Point 130°C to 134°C (14 mmHg)
Flash Point 96°C (204°F)
Refractive Index 1.546
Quantity 25 g
Beilstein 508978
Formula Weight 215.1
Percent Purity 98%
Chemical Name or Material 1-Bromo-3-phenoxypropane
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 209-623-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only