missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-nitrobenzene 98.0+%, TCI America™

Catalog No. B062325G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B062325G 25 g
B0623100G 100 g
B0623500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B062325G Supplier TCI America Supplier No. B062325G
Only null left

Chemical Identifiers

CAS 585-79-5
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.01
MDL Number MFCD00024298
InChI Key FWIROFMBWVMWLB-UHFFFAOYSA-N
Synonym 3-bromonitrobenzene, m-bromonitrobenzene, benzene, 1-bromo-3-nitro, 3-nitrobromobenzene, m-nitrobromobenzene, 1-bromo-3-nitro-benzene, 3-bromo-1-nitrobenzene, ccris 3114, meta-bromonitrobenzene, bromonitrobenzene-3
PubChem CID 11457
IUPAC Name 1-bromo-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(Br)=C1

Specifications

Melting Point 55°C
Color Yellow
Boiling Point 257°C
Quantity 25 g
UN Number 3459
Formula Weight 202.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-3-nitrobenzene
RTECSNumber CY9040500
TSCA Yes