missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-methyl-2-butene (stabilized with Silver chip) 90.0+%, TCI America™

Catalog No. p-7118705
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08445G 5 g
B084425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B08445G Supplier TCI America Supplier No. B08445G
Only null left

Chemical Identifiers

CAS 870-63-3
Molecular Formula C5H9Br
Molecular Weight (g/mol) 149.031
MDL Number MFCD00000242
InChI Key LOYZVRIHVZEDMW-UHFFFAOYSA-N
Synonym 1-bromo-3-methyl-2-butene, 3,3-dimethylallyl bromide, prenyl bromide, 3-methyl-2-butenyl bromide, 2-butene, 1-bromo-3-methyl, dimethylallyl bromide, 1-bromo-3-methyl-but-2-ene, .gamma.,.gamma.-dimethylallyl bromide, 3-methylcrotyl bromide, 3,3-dimethylallylbromide
PubChem CID 70092
IUPAC Name 1-bromo-3-methylbut-2-ene
SMILES CC(=CCBr)C

Specifications

Color Yellow
Boiling Point 135°C
Quantity 5 g
UN Number 2920
Formula Weight 149.03
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-3-methyl-2-butene (stabilized with Silver chip)
EINECSNumber (2)-0067&(2)-2368
TSCA Yes
Recommended Storage Refrigerator