missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-methyl-2-butene, 96%

Catalog No. AC307410100
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC307410100 10 g
AC307410500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC307410100 Supplier Thermo Scientific Chemicals Supplier No. 307410100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 870-63-3 | C5H9Br | 149.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 870-63-3
Molecular Formula C5H9Br
Molecular Weight (g/mol) 149.04
MDL Number MFCD00000242
InChI Key LOYZVRIHVZEDMW-UHFFFAOYSA-N
Synonym 1-bromo-3-methyl-2-butene, 3,3-dimethylallyl bromide, prenyl bromide, 3-methyl-2-butenyl bromide, 2-butene, 1-bromo-3-methyl, dimethylallyl bromide, 1-bromo-3-methyl-but-2-ene, .gamma.,.gamma.-dimethylallyl bromide, 3-methylcrotyl bromide, 3,3-dimethylallylbromide
PubChem CID 70092
IUPAC Name 1-bromo-3-methylbut-2-ene
SMILES CC(=CCBr)C

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Brown or Yellow
Density 1.2700g/mL
Boiling Point 82.0°C to 83.0°C (150.0 mmHg)
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (ex Br) (Argentometry)
Packaging Glass bottle
Linear Formula BrCH2CH=C(CH3)2
Refractive Index 1.4890 to 1.4910
Quantity 10 g
Specific Gravity 1.27
Stabilizer silver wool
Formula Weight 149.04
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1-Bromo-3-methyl-2-butene, 96%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 212-799-5
TSCA TSCA

RUO – Research Use Only