missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-fluorobenzene, 99%

CAS: 1073-06-9 | C6H4BrF | 175 g/mol

Supplier:  Thermo Scientific Chemicals A1085818

 View more versions of this product

Catalog No. AAA1085818


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Bromo-3-fluorobenzene
-8°C
148°C to 150°C
Aromatic
1.5275
50 g
1853969
QDFKKJYEIFBEFC-UHFFFAOYSA-N
1-bromo-3-fluorobenzene
14082
99%
1073-06-9
1.594
46°C (115°F)
C6H4BrF
MFCD00000326
UN1993
3-bromofluorobenzene, m-bromofluorobenzene, m-fluorobromobenzene, benzene, 1-bromo-3-fluoro, 3-bromfluorbenzen, 3-fluorobromobenzene, m-fluorophenyl bromide, 1-fluoro-3-bromobenzene, 3-fluoro bromobenzene, benzene, 1-fluoro-3-bromo
C1=CC(=CC(=C1)Br)F
175
175.01

Chemical Identifiers

1073-06-9
MFCD00000326
3-bromofluorobenzene, m-bromofluorobenzene, m-fluorobromobenzene, benzene, 1-bromo-3-fluoro, 3-bromfluorbenzen, 3-fluorobromobenzene, m-fluorophenyl bromide, 1-fluoro-3-bromobenzene, 3-fluoro bromobenzene, benzene, 1-fluoro-3-bromo
1-bromo-3-fluorobenzene
C6H4BrF
QDFKKJYEIFBEFC-UHFFFAOYSA-N
14082
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H302-H315-H319
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 214-023-0

RTECSNumber : DA1225000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only