missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-3-chloro-5-fluorobenzene 98.0+%, TCI America™
$44.73 - $177.08
Chemical Identifiers
| CAS | 33863-76-2 |
|---|---|
| Molecular Formula | C6H3BrClF |
| Molecular Weight (g/mol) | 209.442 |
| MDL Number | MFCD00070747 |
| InChI Key | GGMDFPMASIXEIR-UHFFFAOYSA-N |
| Synonym | 3-chloro-5-fluorobromobenzene, 1-bromo3-chloro-5-fluorobenzene, 1-bromo-3-chloro-5-fluoro-benzene, benzene, 1-bromo-3-chloro-5-fluoro, 1,3,5-fluorochlorobromobenzene, pubchem2261, acmc-209i3i, ksc494q7t, 3-chloro-5-fluoro-bromobenzene, 1-bromo-3-chloro-5-flurobenzene |
| PubChem CID | 2736223 |
| IUPAC Name | 1-bromo-3-chloro-5-fluorobenzene |
| SMILES | C1=C(C=C(C=C1Cl)Br)F |
Chemical Identifiers
| 33863-76-2 | |
| 209.442 | |
| GGMDFPMASIXEIR-UHFFFAOYSA-N | |
| 2736223 | |
| C1=C(C=C(C=C1Cl)Br)F |
| C6H3BrClF | |
| MFCD00070747 | |
| 3-chloro-5-fluorobromobenzene, 1-bromo3-chloro-5-fluorobenzene, 1-bromo-3-chloro-5-fluoro-benzene, benzene, 1-bromo-3-chloro-5-fluoro, 1,3,5-fluorochlorobromobenzene, pubchem2261, acmc-209i3i, ksc494q7t, 3-chloro-5-fluoro-bromobenzene, 1-bromo-3-chloro-5-flurobenzene | |
| 1-bromo-3-chloro-5-fluorobenzene |
Specifications
| 33863-76-2 | |
| 72°C | |
| MFCD00070747 | |
| 3-chloro-5-fluorobromobenzene, 1-bromo3-chloro-5-fluorobenzene, 1-bromo-3-chloro-5-fluoro-benzene, benzene, 1-bromo-3-chloro-5-fluoro, 1,3,5-fluorochlorobromobenzene, pubchem2261, acmc-209i3i, ksc494q7t, 3-chloro-5-fluoro-bromobenzene, 1-bromo-3-chloro-5-flurobenzene | |
| C1=C(C=C(C=C1Cl)Br)F | |
| 209.442 | |
| 209.44 | |
| Liquid |
| Yellow | |
| C6H3BrClF | |
| 5 g | |
| GGMDFPMASIXEIR-UHFFFAOYSA-N | |
| 1-bromo-3-chloro-5-fluorobenzene | |
| 2736223 | |
| ≥98.0% (GC) | |
| 1-Bromo-3-chloro-5-fluorobenzene |
Safety and Handling
TSCA : No