missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-3,5-dimethyladamantane 98.0+%, TCI America™
$67.38 - $196.36
Chemical Identifiers
| CAS | 941-37-7 |
|---|---|
| Molecular Formula | C12H19Br |
| Molecular Weight (g/mol) | 243.188 |
| MDL Number | MFCD00077197 |
| InChI Key | QUCXLVDIVQWYJR-UHFFFAOYSA-N |
| Synonym | 1-bromo-3,5-dimethyl-adamantane, tricyclo 3.3.1.13,7 decane, 1-bromo-3,5-dimethyl, acmc-209rqd, 3,5-dimethyladamantylbromide, 1-bromo-3,5-dimethyltricyclo 3.3.1.13,7 decane, 1-bromo-3,5dimethyladamantane, 3,5-dimethyl-1-bromoadamantane, 1-bromo-3,5-dimethyl adamantane, 1-bromanyl-3,5-dimethyl-adamantane, 1-?bromo-?3,5-?dimethyladamantane |
| PubChem CID | 98317 |
| IUPAC Name | 1-bromo-3,5-dimethyladamantane |
| SMILES | CC12CC3CC(C1)(CC(C3)(C2)Br)C |
Chemical Identifiers
| 941-37-7 | |
| 243.188 | |
| QUCXLVDIVQWYJR-UHFFFAOYSA-N | |
| 98317 | |
| CC12CC3CC(C1)(CC(C3)(C2)Br)C |
| C12H19Br | |
| MFCD00077197 | |
| 1-bromo-3,5-dimethyl-adamantane, tricyclo 3.3.1.13,7 decane, 1-bromo-3,5-dimethyl, acmc-209rqd, 3,5-dimethyladamantylbromide, 1-bromo-3,5-dimethyltricyclo 3.3.1.13,7 decane, 1-bromo-3,5dimethyladamantane, 3,5-dimethyl-1-bromoadamantane, 1-bromo-3,5-dimethyl adamantane, 1-bromanyl-3,5-dimethyl-adamantane, 1-?bromo-?3,5-?dimethyladamantane | |
| 1-bromo-3,5-dimethyladamantane |
Specifications
| 941-37-7 | |
| 273°C | |
| MFCD00077197 | |
| 1-bromo-3,5-dimethyl-adamantane, tricyclo 3.3.1.13,7 decane, 1-bromo-3,5-dimethyl, acmc-209rqd, 3,5-dimethyladamantylbromide, 1-bromo-3,5-dimethyltricyclo 3.3.1.13,7 decane, 1-bromo-3,5dimethyladamantane, 3,5-dimethyl-1-bromoadamantane, 1-bromo-3,5-dimethyl adamantane, 1-bromanyl-3,5-dimethyl-adamantane, 1-?bromo-?3,5-?dimethyladamantane | |
| CC12CC3CC(C1)(CC(C3)(C2)Br)C | |
| 243.188 | |
| 243.19 | |
| Liquid |
| Yellow | |
| C12H19Br | |
| 5 g | |
| QUCXLVDIVQWYJR-UHFFFAOYSA-N | |
| 1-bromo-3,5-dimethyladamantane | |
| 98317 | |
| ≥98.0% (GC) | |
| 1-Bromo-3,5-dimethyladamantane |
Safety and Handling
TSCA : No