missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,5-difluorobenzene 98.0+%, TCI America™

Catalog No. p-7119248
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B176425G 25 g
B1764250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B176425G Supplier TCI America Supplier No. B176425G
Only null left

Chemical Identifiers

CAS 461-96-1
Molecular Formula C6H3BrF2
Molecular Weight (g/mol) 192.99
MDL Number MFCD00010305
InChI Key JHLKSIOJYMGSMB-UHFFFAOYSA-N
Synonym 3,5-difluorobromobenzene, 1-bromo-3,5-difluoro-benzene, 1-bromo-3,5-difluoro benzene, benzene, 1-bromo-3,5-difluoro, 3,5-difluoro-bromobenzene, pubchem1042, acmc-1ae0k, 3,5-difluoro bromobenzene, bromo-3,5-difluorobenzene, 3,5-difluorophenyl bromide
PubChem CID 136313
IUPAC Name 1-bromo-3,5-difluorobenzene
SMILES FC1=CC(Br)=CC(F)=C1

Specifications

Color Yellow
Boiling Point 140°C
Quantity 25 g
UN Number 1993
Formula Weight 192.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-3,5-difluorobenzene