missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,5-dichlorobenzene, 97%

Catalog No. AAB2549206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2549206 5 g
AAB2549214 25 g
AAB2549222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2549206 Supplier Thermo Scientific Chemicals Supplier No. B2549206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 19752-55-7 | C6H3BrCl2 | 225.894 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19752-55-7
Molecular Formula C6H3BrCl2
Molecular Weight (g/mol) 225.894
MDL Number MFCD00000584
InChI Key DZHFFMWJXJBBRG-UHFFFAOYSA-N
Synonym 3,5-dichlorobromobenzene, benzene, 1-bromo-3,5-dichloro, 1-bromo-3,5-dichloro-benzene, 3,5-dichloro bromobenzene, 3,5-dichloro-1-bromobenzene, attercop-chm at113311, maybridge1_000881, pubchem3612, acmc-209f1a
PubChem CID 29766
IUPAC Name 1-bromo-3,5-dichlorobenzene
SMILES C1=C(C=C(C=C1Cl)Br)Cl

Specifications

Melting Point 73°C to 77°C
Boiling Point 232°C
Quantity 5 g
Beilstein 1931524
Formula Weight 225.91
Percent Purity 97%
Chemical Name or Material 1-Bromo-3,5-dichlorobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 243-270-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only