missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,5-bis(tert-butylthio)benzene 98.0+%, TCI America™

Catalog No. B26581G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B26581G Supplier TCI America Supplier No. B26581G
Only null left

Chemical Identifiers

CAS 795274-44-1
Molecular Formula C14H21BrS2
Molecular Weight (g/mol) 333.35
MDL Number MFCD08276293
InChI Key VLQVYTYMAXNJGM-UHFFFAOYSA-N
Synonym 3,5-Bis(tert-butylthio)-1-bromobenzene
PubChem CID 11739095
IUPAC Name 1-bromo-3,5-bis(tert-butylsulfanyl)benzene
SMILES CC(C)(C)SC1=CC(SC(C)(C)C)=CC(Br)=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 333.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-3,5-bis(tert-butylthio)benzene