missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,4-difluorobenzene 98.0+%, TCI America™

Catalog No. B152325G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B152325G 25 g
B1523250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B152325G Supplier TCI America Supplier No. B152325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 348-61-8
Molecular Formula C6H3BrF2
Molecular Weight (g/mol) 192.99
MDL Number MFCD00000304
InChI Key YMQPKONILWWJQG-UHFFFAOYSA-N
Synonym 1-bromo-3,4-difluorobenzene, 3,4-difluorobromobenzene, benzene, 4-bromo-1,2-difluoro, 3,4-difluorobromebenzene, 4-bromo-1,2-difluoro-benzene, 3,4-difluoro-1-bromobenzene, paragos 390209, fr bf de, pubchem2303, 3,4-difluorophenylbromide
PubChem CID 67675
IUPAC Name 4-bromo-1,2-difluorobenzene
SMILES FC1=CC=C(Br)C=C1F

Specifications

Color Yellow
Boiling Point 151°C
Quantity 25 g
UN Number 1993
Formula Weight 192.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-3,4-difluorobenzene
EINECSNumber (3)-4148
TSCA No