missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,4,5-trimethoxybenzene, 97%

Numéro de catalogue. p-7077716
Modifier l'affichage
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AC458130050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AC458130050 Fournisseur Thermo Scientific Chemicals Code fournisseur 458130050
Il en reste null

CAS: 2675-79-8 | C9H11BrO3 | 247.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 2675-79-8
Molecular Formula C9H11BrO3
Molecular Weight (g/mol) 247.09
MDL Number MFCD00017169
InChI Key XAOOZMATJDXDQJ-UHFFFAOYSA-N
Synonym 1-bromo-3,4,5-trimethoxybenzene, 3,4,5-trimethoxybromobenzene, bromotrimethoxybenzene, benzene, 5-bromo-1,2,3-trimethoxy, 5-bromo-1,2,3-trimethoxy-benzene, pubchem10980, 3,4,5-trimethoxybromoben, ksc490o1d, 3,4,5-trimethoxyphenylbromide, 3,4,5-triemethoxyphenylbromide
PubChem CID 75885
IUPAC Name 5-bromo-1,2,3-trimethoxybenzene
SMILES COC1=CC(Br)=CC(OC)=C1OC

Spécifications

Melting Point 78.0°C to 82.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 247.09
Percent Purity 97%
Chemical Name or Material 1-Bromo-3,4,5-trimethoxybenzene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
EINECSNumber 220-223-9

RUO – Research Use Only