missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,4,5-trifluorobenzene 98.0+%, TCI America™

Catalog No. B17655G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17655G 5 g
B176525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17655G Supplier TCI America Supplier No. B17655G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 138526-69-9
Molecular Formula C6H2BrF3
Molecular Weight (g/mol) 210.981
MDL Number MFCD00070738
InChI Key HKJCELUUIFFSIN-UHFFFAOYSA-N
Synonym 1-bromo-3,4,5-trifluorobenzene, 3,4,5-trifluorobromobenzene, 1,2,3-trifluoro-5-bromobenzene, 5-bromo-1,2,3-trifluoro-benzene, benzene, 5-bromo-1,2,3-trifluoro, pubchem2304, acmc-1c02k, ksc174s3f, 3,4,5-trifluoro-bromobenzene, 3,4,5-trifluorophenyl bromide
PubChem CID 611409
IUPAC Name 5-bromo-1,2,3-trifluorobenzene
SMILES C1=C(C=C(C(=C1F)F)F)Br

Specifications

Color Yellow
Boiling Point 43°C
Quantity 5 g
UN Number 1993
Formula Weight 210.98
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-3,4,5-trifluorobenzene
EINECSNumber (3)-4279
TSCA No