missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,3,3-trifluoroacetone, 97%

Catalog No. p-7022259
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1494806 5 g
AAA1494814 25 g
AAA1494822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1494806 Supplier Thermo Scientific Chemicals Supplier No. A1494806
Only null left

CAS: 431-35-6 | C3H2BrF3O | 190.95 g/mol

1-Bromo-3,3,3-trifluoroacetone is used in the preparation of 3-nonylthio-1,1,1-trifluoropropan-2-one and perfluoroalkylated trans-allylic alcohols. It is involved in the synthesis of cyclic tetrapeptides. Further, it reacts with potassium enolate of ethyl 4,4,4-trifluoroacetoacetate to produce ethyl 2,4-bis(trifluoromethyl)-4-hydroxydihydro-3-furoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3,3,3-trifluoroacetone is used in the preparation of 3-nonylthio-1,1,1-trifluoropropan-2-one and perfluoroalkylated trans-allylic alcohols. It is involved in the synthesis of cyclic tetrapeptides. Further, it reacts with potassium enolate of ethyl 4,4,4-trifluoroacetoacetate to produce ethyl 2,4-bis(trifluoromethyl)-4-hydroxydihydro-3-furoate.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and bases.

Chemical Identifiers

CAS 431-35-6
Molecular Formula C3H2BrF3O
Molecular Weight (g/mol) 190.95
MDL Number MFCD00039237
InChI Key ONZQYZKCUHFORE-UHFFFAOYSA-N
Synonym 3-bromo-1,1,1-trifluoroacetone, 1-bromo-3,3,3-trifluoroacetone, bromotrifluoroacetone, 3-bromo-1,1,1-trifluoro-2-propanone, 3-bromo-1,1,1-trifluoropropanone, 1-bromo-3,3,3-trifluoro-2-propanone, 2-propanone, 3-bromo-1,1,1-trifluoro, 1,1,1-trifluoro-3-bromopropanone, 3-bromo-1,1,1-trifluoro-propan-2-one, bromotrifloroacetone
PubChem CID 79008
IUPAC Name 3-bromo-1,1,1-trifluoropropan-2-one
SMILES FC(F)(F)C(=O)CBr

Specifications

Melting Point <-80°C
Density 1.822
Boiling Point 85°C to 87°C
Flash Point −2°C (28°F)
Odor Pungent
Refractive Index 1.376
Quantity 5 g
UN Number UN2924
Beilstein 1703387
Solubility Information Immiscible with water.
Formula Weight 190.95
Percent Purity 97%
Chemical Name or Material 1-Bromo-3,3,3-trifluoroacetone
Show More Show Less
Hazard Category H225-H314
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 207-071-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only