missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-(trifluoromethylthio)benzene, 97%

Catalog No. AAH5057603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5057603 1 g
AAH5057606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5057603 Supplier Thermo Scientific Chemicals Supplier No. H5057603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1644-72-0 | C7H4BrF3S | 257.07 g/mol

1-Bromo-2-(trifluoromethylthio)benzene is used as organic chemical synthesis intermediate. It is also a useful halogenated compound for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-(trifluoromethylthio)benzene is used as organic chemical synthesis intermediate. It is also a useful halogenated compound for proteomics research.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 1644-72-0
Molecular Formula C7H4BrF3S
Molecular Weight (g/mol) 257.07
MDL Number MFCD00236347
InChI Key IWRJVJJQWGZKMT-UHFFFAOYSA-N
Synonym 2-trifluoromethylthio bromobenzene, 1-bromo-2-trifluoromethylthio benzene, 1-bromo-2-trifluoromethyl sulfanyl benzene, 2-bromophenyl trifluoromethyl sulfane, 2-bromophenyl trifluoromethyl sulphide, pubchem20490, 1-bromo-2-trifluoromethylsulfanyl benzene, 1-trifluoromethylthio-2-bromobenzene, 2-bromo-1-trifluoromethylthio benzene, 2-bromophenyl trifluoromethyl sulfide
PubChem CID 2779338
IUPAC Name 1-bromo-2-(trifluoromethylsulfanyl)benzene
SMILES FC(F)(F)SC1=CC=CC=C1Br

Specifications

Boiling Point 92°C to 94°C (20 mmHg)
Refractive Index 1.5125
Quantity 1 g
Formula Weight 257.07
Percent Purity 97%
Chemical Name or Material 1-Bromo-2-(trifluoromethylthio)benzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only