missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2-phenylnaphthalene 98.0+%, TCI America™
$205.05 - $615.14
Chemical Identifiers
| CAS | 22082-93-5 |
|---|---|
| Molecular Formula | C16H11Br |
| Molecular Weight (g/mol) | 283.168 |
| InChI Key | ANKXTSUKPPHQJR-UHFFFAOYSA-N |
| PubChem CID | 10956972 |
| IUPAC Name | 1-bromo-2-phenylnaphthalene |
| SMILES | C1=CC=C(C=C1)C2=C(C3=CC=CC=C3C=C2)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B4636200MG
|
TCI America
B4636200MG |
200 mg |
Each for $205.05
|
|
|||||
|
B46361G
|
TCI America
B46361G |
1 g |
Each for $615.14
|
|
|||||
Chemical Identifiers
| 22082-93-5 | |
| 283.168 | |
| 10956972 | |
| C1=CC=C(C=C1)C2=C(C3=CC=CC=C3C=C2)Br |
| C16H11Br | |
| ANKXTSUKPPHQJR-UHFFFAOYSA-N | |
| 1-bromo-2-phenylnaphthalene |
Specifications
| 22082-93-5 | |
| White-Yellow | |
| C16H11Br | |
| ANKXTSUKPPHQJR-UHFFFAOYSA-N | |
| 1-bromo-2-phenylnaphthalene | |
| 10956972 | |
| ≥98.0% (GC) | |
| 1-Bromo-2-phenylnaphthalene |
| 65°C | |
| 170°C | |
| 200 mg | |
| C1=CC=C(C=C1)C2=C(C3=CC=CC=C3C=C2)Br | |
| 283.168 | |
| 283.17 | |
| Crystalline Powder |
Safety and Handling
TSCA : No