missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-methylpropane, 98+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 168830025

Catalog No. AC168830025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1-Bromo-2-methylpropane
1.2600g/mL
18°C
98% min. (GC)
C4H9Br
(CH3)2CHCH2Br
01,126
15,518
Solubility in water: 0.51g/L (18°C). Other solubilities: soluble in most common organic solvents
CC(C)CBr
137.02
137.02
78-77-3
90.0°C to 92.0°C
Authentic
Glass bottle
1.4346 to 1.4366
2.5L
1.26
isobutyl bromide, iso-butyl bromide, propane, 1-bromo-2-methyl, bromoisobutane, i-butyl bromide, 1-bromo-2-methyl-propane, unii-5oec0bw987, ccris 349, 1-bromo-2-methyl propane, bromoacetaldehydedimethylacetal
HLVFKOKELQSXIQ-UHFFFAOYSA-N
1-bromo-2-methylpropane
6555
98+%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protect

GHS Signal Word: Danger

EINECSNumber : 201-141-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only