Learn More
1-Bromo-2-methoxynaphthalene, 97%
CAS: 3401-47-6 | C11H9BrO | 237.10 g/mol
Supplier: Thermo Scientific Chemicals A1892606
Description
1-Bromo-2-methoxy-naphthalene is a useful synthetic intermediate. It is used as a reagent to synthesize a catalyst for highly enantioselective aziridination of styrene derivatives. It can also be used to prepare biaryls or biheterocycles by palladium-catalyzed Ullmann coupling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromo-2-methoxy-naphthalene is a useful synthetic intermediate. It is used as a reagent to synthesize a catalyst for highly enantioselective aziridination of styrene derivatives. It can also be used to prepare biaryls or biheterocycles by palladium-catalyzed Ullmann coupling.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| 1-Bromo-2-methoxynaphthalene | |
| 79°C to 80°C | |
| MFCD00055374 | |
| 2045600 | |
| Slightly soluble in water. | |
| COC1=CC=C2C=CC=CC2=C1Br | |
| 237.10 | |
| 237.1 |
| 3401-47-6 | |
| C11H9BrO | |
| 5 g | |
| 1-bromo-2-methoxy-naphthalene, naphthalene, 1-bromo-2-methoxy, acmc-1cjky, maybridge3_005134, 2-methoxy-1-bromonaphthalene, 1-bromo-2-methoxy-naphtalene | |
| XNIGURFWNPLWJM-UHFFFAOYSA-N | |
| 1-bromo-2-methoxynaphthalene | |
| 72860 | |
| 97% |
Chemical Identifiers
| 3401-47-6 | |
| 237.10 | |
| XNIGURFWNPLWJM-UHFFFAOYSA-N | |
| 72860 | |
| COC1=CC=C2C=CC=CC2=C1Br |
| C11H9BrO | |
| MFCD00055374 | |
| 1-bromo-2-methoxy-naphthalene, naphthalene, 1-bromo-2-methoxy, acmc-1cjky, maybridge3_005134, 2-methoxy-1-bromonaphthalene, 1-bromo-2-methoxy-naphtalene | |
| 1-bromo-2-methoxynaphthalene |
Safety and Handling
EINECSNumber : 222-272-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only