accessibility menu, dialog, popup

UserName

CAS RN 3401-47-6

IUPAC Name: 1-bromo-2-methoxynaphthalene
Synonyms: 1-bromo-2-methoxynaphthalene
Molecular Weight (g/mol): 237.1
Molecular Formula: C11H9BrO
InChi Key: XNIGURFWNPLWJM-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=CC=CC2=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)