missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-methoxynaphthalene, 97%

Catalog No. AAA1892622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1892622 100 g
AAA1892606 5 g
AAA1892614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1892622 Supplier Thermo Scientific Chemicals Supplier No. A1892622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3401-47-6 | C11H9BrO | 237.10 g/mol

1-Bromo-2-methoxy-naphthalene is a useful synthetic intermediate. It is used as a reagent to synthesize a catalyst for highly enantioselective aziridination of styrene derivatives. It can also be used to prepare biaryls or biheterocycles by palladium-catalyzed Ullmann coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-methoxy-naphthalene is a useful synthetic intermediate. It is used as a reagent to synthesize a catalyst for highly enantioselective aziridination of styrene derivatives. It can also be used to prepare biaryls or biheterocycles by palladium-catalyzed Ullmann coupling.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 3401-47-6
Molecular Formula C11H9BrO
Molecular Weight (g/mol) 237.10
MDL Number MFCD00055374
InChI Key XNIGURFWNPLWJM-UHFFFAOYSA-N
Synonym 1-bromo-2-methoxy-naphthalene, naphthalene, 1-bromo-2-methoxy, acmc-1cjky, maybridge3_005134, 2-methoxy-1-bromonaphthalene, 1-bromo-2-methoxy-naphtalene
PubChem CID 72860
IUPAC Name 1-bromo-2-methoxynaphthalene
SMILES COC1=CC=C2C=CC=CC2=C1Br

Specifications

Melting Point 79°C to 80°C
Quantity 100 g
Beilstein 2045600
Solubility Information Slightly soluble in water.
Formula Weight 237.1
Percent Purity 97%
Chemical Name or Material 1-Bromo-2-methoxynaphthalene
EINECSNumber 222-272-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only