missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene, 98%

Catalog No. p-7031536
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3265003 1 g
AAH3265006 5 g
AAH3265014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3265003 Supplier Thermo Scientific Chemicals Supplier No. H3265003

CAS: 286932-57-8 | C7H3BrF4O | 258.998 g/mol

1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene used as solvents. Mostly employed in chemical processing, extraction and separation of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene used as solvents. Mostly employed in chemical processing, extraction and separation of chemicals.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 286932-57-8
Molecular Formula C7H3BrF4O
Molecular Weight (g/mol) 258.998
MDL Number MFCD02181186
InChI Key VVVFPEJORKPEST-UHFFFAOYSA-N
Synonym 2-bromo-1-fluoro-4-trifluoromethoxy benzene, 1-bromo-2-fluoro-5-trifluoromethoxy benzene, 3-bromo-4-fluoro trifluoromethoxy benzene, 3-bromo-4-fluorotrifluoroanisole, 2-fluoro-5-trifluoromethoxy-bromobenzene, 3-bromo-4-fluorophenyl trifluoromethyl ether, 3-bromo-4-fluoro-alpha,alpha,alpha-trifluoroanisole, 3-bromo-4-fluorotrifluoromethoxybenzene, pubchem12363, acmc-1cocq
PubChem CID 25067359
IUPAC Name 2-bromo-1-fluoro-4-(trifluoromethoxy)benzene
SMILES C1=CC(=C(C=C1OC(F)(F)F)Br)F

Specifications

Quantity 1 g
Formula Weight 259
Percent Purity 98%
Chemical Name or Material 1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only