missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2-fluoro-4-nitrobenzene 95.0+%, TCI America™
$93.21 - $285.21
Chemical Identifiers
| CAS | 185331-69-5 |
|---|---|
| Molecular Formula | C6H3BrFNO2 |
| Molecular Weight (g/mol) | 220.00 |
| MDL Number | MFCD05865088 |
| InChI Key | AZYQMHQOHNGPRR-UHFFFAOYSA-N |
| Synonym | 4-bromo-3-fluoronitrobenzene, 3-fluoro-4-bromonitrobenzene, benzene, 1-bromo-2-fluoro-4-nitro, pubchem1045, acmc-1byt6, ksc495e5r, 1-bromo-2-fluoro-4-nitro-benzene, 4-bromo-3-fluoronitrobenzene; 3-fluoro-4-bromonitrobenzene |
| PubChem CID | 2756994 |
| IUPAC Name | 1-bromo-2-fluoro-4-nitrobenzene |
| SMILES | [O-][N+](=O)C1=CC(F)=C(Br)C=C1 |
Chemical Identifiers
| 185331-69-5 | |
| 220.00 | |
| AZYQMHQOHNGPRR-UHFFFAOYSA-N | |
| 2756994 | |
| [O-][N+](=O)C1=CC(F)=C(Br)C=C1 |
| C6H3BrFNO2 | |
| MFCD05865088 | |
| 4-bromo-3-fluoronitrobenzene, 3-fluoro-4-bromonitrobenzene, benzene, 1-bromo-2-fluoro-4-nitro, pubchem1045, acmc-1byt6, ksc495e5r, 1-bromo-2-fluoro-4-nitro-benzene, 4-bromo-3-fluoronitrobenzene; 3-fluoro-4-bromonitrobenzene | |
| 1-bromo-2-fluoro-4-nitrobenzene |
Specifications
| 185331-69-5 | |
| C6H3BrFNO2 | |
| 5 g | |
| AZYQMHQOHNGPRR-UHFFFAOYSA-N | |
| 1-bromo-2-fluoro-4-nitrobenzene | |
| 2756994 | |
| ≥95.0% (GC) | |
| 1-Bromo-2-fluoro-4-nitrobenzene |
| 91°C | |
| MFCD05865088 | |
| 4-bromo-3-fluoronitrobenzene, 3-fluoro-4-bromonitrobenzene, benzene, 1-bromo-2-fluoro-4-nitro, pubchem1045, acmc-1byt6, ksc495e5r, 1-bromo-2-fluoro-4-nitro-benzene, 4-bromo-3-fluoronitrobenzene; 3-fluoro-4-bromonitrobenzene | |
| [O-][N+](=O)C1=CC(F)=C(Br)C=C1 | |
| 220.00 | |
| 220.00 | |
| Crystalline Powder |
Safety and Handling
TSCA : No