missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-fluoro-4-nitrobenzene 95.0+%, TCI America™

Catalog No. B30605G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30605G 5 g
B306025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30605G Supplier TCI America Supplier No. B30605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 185331-69-5
Molecular Formula C6H3BrFNO2
Molecular Weight (g/mol) 220.00
MDL Number MFCD05865088
InChI Key AZYQMHQOHNGPRR-UHFFFAOYSA-N
Synonym 4-bromo-3-fluoronitrobenzene, 3-fluoro-4-bromonitrobenzene, benzene, 1-bromo-2-fluoro-4-nitro, pubchem1045, acmc-1byt6, ksc495e5r, 1-bromo-2-fluoro-4-nitro-benzene, 4-bromo-3-fluoronitrobenzene; 3-fluoro-4-bromonitrobenzene
PubChem CID 2756994
IUPAC Name 1-bromo-2-fluoro-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC(F)=C(Br)C=C1

Specifications

Melting Point 91°C
Quantity 5 g
Formula Weight 220.00
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-2-fluoro-4-nitrobenzene