missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-fluoro-4-nitrobenzene, 98%

Catalog No. AAH6149414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6149414 25 g
AAH6149422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6149414 Supplier Thermo Scientific Chemicals Supplier No. H6149414

CAS: 185331-69-5 | C6H3BrFNO2 | 220.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 185331-69-5
Molecular Formula C6H3BrFNO2
Molecular Weight (g/mol) 220.00
MDL Number MFCD05865088
InChI Key AZYQMHQOHNGPRR-UHFFFAOYSA-N
Synonym 4-bromo-3-fluoronitrobenzene, 3-fluoro-4-bromonitrobenzene, benzene, 1-bromo-2-fluoro-4-nitro, pubchem1045, acmc-1byt6, ksc495e5r, 1-bromo-2-fluoro-4-nitro-benzene, 4-bromo-3-fluoronitrobenzene; 3-fluoro-4-bromonitrobenzene
PubChem CID 2756994
IUPAC Name 1-bromo-2-fluoro-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC(F)=C(Br)C=C1

Specifications

Melting Point 88°C to 94°C
Quantity 25 g
Formula Weight 220
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1-Bromo-2-fluoro-4-nitrobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only