missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2-ethylbutane (stabilized with Copper chip) 97.0+%, TCI America™
$123.03 - $350.44
Chemical Identifiers
| CAS | 3814-34-4 |
|---|---|
| Molecular Formula | C6H13Br |
| Molecular Weight (g/mol) | 165.074 |
| MDL Number | MFCD00000219 |
| InChI Key | KKGUMGWNFARLSL-UHFFFAOYSA-N |
| Synonym | 1-bromo-2-ethylbutane, 3-bromomethyl pentane, 2-ethylbutyl bromide, pentane, 3-bromomethyl, 3-bromomethyl-pentane, butane, 1-bromo-2-ethyl, 2-ethylbutylbromide, bromo-2-ethylbutane, 2-ethyl butyl bromide, 2-ethyl-1-bromobutane |
| PubChem CID | 77432 |
| IUPAC Name | 3-(bromomethyl)pentane |
| SMILES | CCC(CC)CBr |
Chemical Identifiers
| 3814-34-4 | |
| 165.074 | |
| KKGUMGWNFARLSL-UHFFFAOYSA-N | |
| 77432 | |
| CCC(CC)CBr |
| C6H13Br | |
| MFCD00000219 | |
| 1-bromo-2-ethylbutane, 3-bromomethyl pentane, 2-ethylbutyl bromide, pentane, 3-bromomethyl, 3-bromomethyl-pentane, butane, 1-bromo-2-ethyl, 2-ethylbutylbromide, bromo-2-ethylbutane, 2-ethyl butyl bromide, 2-ethyl-1-bromobutane | |
| 3-(bromomethyl)pentane |
Specifications
| 3814-34-4 | |
| 145°C | |
| MFCD00000219 | |
| 1993 | |
| KKGUMGWNFARLSL-UHFFFAOYSA-N | |
| 3-(bromomethyl)pentane | |
| 77432 | |
| ≥97.0% (GC) | |
| 1-Bromo-2-ethylbutane (stabilized with Copper chip) |
| Yellow | |
| C6H13Br | |
| 5 g | |
| 1-bromo-2-ethylbutane, 3-bromomethyl pentane, 2-ethylbutyl bromide, pentane, 3-bromomethyl, 3-bromomethyl-pentane, butane, 1-bromo-2-ethyl, 2-ethylbutylbromide, bromo-2-ethylbutane, 2-ethyl butyl bromide, 2-ethyl-1-bromobutane | |
| CCC(CC)CBr | |
| 165.074 | |
| 165.07 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0067
TSCA : Yes