Learn More
1-Bromo-2-ethylbutane, 97%
CAS: 3814-34-4 | C6H13Br | 165.074 g/mol
$44.66 - $149.00
Chemical Identifiers
| CAS | 3814-34-4 |
|---|---|
| Molecular Formula | C6H13Br |
| Molecular Weight (g/mol) | 165.074 |
| MDL Number | MFCD00000219 |
| InChI Key | KKGUMGWNFARLSL-UHFFFAOYSA-N |
| Synonym | 1-bromo-2-ethylbutane, 3-bromomethyl pentane, 2-ethylbutyl bromide, pentane, 3-bromomethyl, 3-bromomethyl-pentane, butane, 1-bromo-2-ethyl, 2-ethylbutylbromide, bromo-2-ethylbutane, 2-ethyl butyl bromide, 2-ethyl-1-bromobutane |
| PubChem CID | 77432 |
| IUPAC Name | 3-(bromomethyl)pentane |
| SMILES | CCC(CC)CBr |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH26445AA
|
Thermo Scientific Chemicals
H26445AA |
5 mL |
Each for $44.66
|
|
|||||
|
AAH26445AC
|
Thermo Scientific Chemicals
H26445AC |
25 mL |
Each for $149.00
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3814-34-4 | |
| 165.074 | |
| KKGUMGWNFARLSL-UHFFFAOYSA-N | |
| 77432 | |
| CCC(CC)CBr |
| C6H13Br | |
| MFCD00000219 | |
| 1-bromo-2-ethylbutane, 3-bromomethyl pentane, 2-ethylbutyl bromide, pentane, 3-bromomethyl, 3-bromomethyl-pentane, butane, 1-bromo-2-ethyl, 2-ethylbutylbromide, bromo-2-ethylbutane, 2-ethyl butyl bromide, 2-ethyl-1-bromobutane | |
| 3-(bromomethyl)pentane |
Specifications
| 3814-34-4 | |
| 145°C | |
| Sweet | |
| 1.4515 | |
| 5 mL | |
| 1-bromo-2-ethylbutane, 3-bromomethyl pentane, 2-ethylbutyl bromide, pentane, 3-bromomethyl, 3-bromomethyl-pentane, butane, 1-bromo-2-ethyl, 2-ethylbutylbromide, bromo-2-ethylbutane, 2-ethyl butyl bromide, 2-ethyl-1-bromobutane | |
| CCC(CC)CBr | |
| 165.074 | |
| 165.07 | |
| 1-Bromo-2-ethylbutane |
| 1.179 | |
| 36°C (96°F) | |
| C6H13Br | |
| MFCD00000219 | |
| UN1993 | |
| KKGUMGWNFARLSL-UHFFFAOYSA-N | |
| 3-(bromomethyl)pentane | |
| 77432 | |
| 97% |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 223-302-6
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only