missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-ethylbenzene 98.0+%, TCI America™

Catalog No. p-7119050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14285G 5 g
B142825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14285G Supplier TCI America Supplier No. B14285G
Only null left

Chemical Identifiers

CAS 1973-22-4
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000077
InChI Key HVRUGFJYCAFAAN-UHFFFAOYSA-N
PubChem CID 16089
IUPAC Name 1-bromo-2-ethylbenzene
SMILES CCC1=CC=CC=C1Br

Specifications

Color Yellow
Boiling Point 199°C
Quantity 5 g
Synonym o-bromoethylbenzene, benzene, 1-bromo-2-ethyl, 2-ethylbromobenzene, 1-ethyl-2-bromobenzene, 2-bromo-1-ethylbenzene, 1-bromo-2-ethyl-benzene, benzene,1-bromo-2-ethyl, o-ethylbromobenzene, 2-bromophenyl ethane, bromo-2-ethylbenzene
Formula Weight 185.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-ethylbenzene