missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-ethylbenzene, 98%

Catalog No. p-7027014
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2344906 5 g
AAB2344914 25 g
AAB2344922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2344906 Supplier Thermo Scientific Chemicals Supplier No. B2344906
Only null left

CAS: 1973-22-4 | C8H9Br | 185.064 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1973-22-4
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000077
InChI Key HVRUGFJYCAFAAN-UHFFFAOYSA-N
PubChem CID 16089
IUPAC Name 1-bromo-2-ethylbenzene
SMILES CCC1=CC=CC=C1Br

Specifications

Melting Point -68°C
Density 1.338
Boiling Point 199°C to 200°C
Flash Point 71°C (159°F)
Refractive Index 1.5485
Quantity 5 g
Beilstein 1929735
Synonym o-bromoethylbenzene, benzene, 1-bromo-2-ethyl, 2-ethylbromobenzene, 1-ethyl-2-bromobenzene, 2-bromo-1-ethylbenzene, 1-bromo-2-ethyl-benzene, benzene,1-bromo-2-ethyl, o-ethylbromobenzene, 2-bromophenyl ethane, bromo-2-ethylbenzene
Formula Weight 185.06
Percent Purity 98%
Chemical Name or Material 1-Bromo-2-ethylbenzene
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 217-823-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only