missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-cyclohexylbenzene, 97%

Catalog No. AAL1805903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1805903 1 g
AAL1805906 5 g
AAL1805914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1805903 Supplier Thermo Scientific Chemicals Supplier No. L1805903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 59734-92-8 | C12H15Br | 239.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59734-92-8
Molecular Formula C12H15Br
Molecular Weight (g/mol) 239.16
MDL Number MFCD02093987
InChI Key BXMDAGGQRIDSPA-UHFFFAOYSA-N
Synonym 1-bromo-2-cyclohexyl-benzene, benzene, 1-bromo-2-cyclohexyl, acmc-20aoxa, l-bromo-2-cyclohexylbenzene, 2-bromo-1-cyclohexylbenzene
PubChem CID 10944502
IUPAC Name 1-bromo-2-cyclohexylbenzene
SMILES BrC1=CC=CC=C1C1CCCCC1

Specifications

Density 1.295
Boiling Point 79°C to 83°C (0.01 mmHg)
Refractive Index 1.5605
Quantity 1 g
Beilstein 2365764
Formula Weight 239.16
Percent Purity 97%
Chemical Name or Material 1-Bromo-2-cyclohexylbenzene
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only