missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2-chloroethane 98.0+%, TCI America™
Supplier: TCI America B0572100G
Specifications
| 1-Bromo-2-chloroethane | |
| Colorless | |
| C2H4BrCl | |
| 100 g | |
| ethylene chlorobromide, 2-chloroethyl bromide, 1-chloro-2-bromoethane, 2-bromoethyl chloride, ethane, 1-bromo-2-chloro, 2-bromo-1-chloroethane, s-chlorobromoethane, 1,2-bromochloroethane, beta-chloroethyl bromide, sym-chlorobromoethane | |
| C(CBr)Cl | |
| 143.408 | |
| CHEBI:19032 | |
| ≥98.0% (GC) |
| 107-04-0 | |
| 106°C | |
| MFCD00000962 | |
| 2810 | |
| IBYHHJPAARCAIE-UHFFFAOYSA-N | |
| 1-bromo-2-chloroethane | |
| 7849 | |
| 143.41 | |
| Liquid |
Chemical Identifiers
| 107-04-0 | |
| 143.408 | |
| IBYHHJPAARCAIE-UHFFFAOYSA-N | |
| 7849 | |
| 1-bromo-2-chloroethane |
| C2H4BrCl | |
| MFCD00000962 | |
| ethylene chlorobromide, 2-chloroethyl bromide, 1-chloro-2-bromoethane, 2-bromoethyl chloride, ethane, 1-bromo-2-chloro, 2-bromo-1-chloroethane, s-chlorobromoethane, 1,2-bromochloroethane, beta-chloroethyl bromide, sym-chlorobromoethane | |
| CHEBI:19032 | |
| C(CBr)Cl |
Safety and Handling
EINECSNumber : (2)-3351
RTECSNumber : KH6500000
TSCA : Yes