missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-chloroethane 98.0+%, TCI America™

Catalog No. p-7118535
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B057225G 25 g
B0572100G 100 g
B0572500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B057225G Supplier TCI America Supplier No. B057225G
Only null left

Chemical Identifiers

CAS 107-04-0
Molecular Formula C2H4BrCl
Molecular Weight (g/mol) 143.408
MDL Number MFCD00000962
InChI Key IBYHHJPAARCAIE-UHFFFAOYSA-N
Synonym ethylene chlorobromide, 2-chloroethyl bromide, 1-chloro-2-bromoethane, 2-bromoethyl chloride, ethane, 1-bromo-2-chloro, 2-bromo-1-chloroethane, s-chlorobromoethane, 1,2-bromochloroethane, beta-chloroethyl bromide, sym-chlorobromoethane
PubChem CID 7849
ChEBI CHEBI:19032
IUPAC Name 1-bromo-2-chloroethane
SMILES C(CBr)Cl

Specifications

Color Colorless
Boiling Point 106°C
Quantity 25 g
UN Number 2810
Formula Weight 143.41
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-chloroethane
EINECSNumber (2)-3351
RTECSNumber KH6500000
TSCA Yes