missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-chloro-4-nitrobenzene, 98%

Catalog No. aal0757309
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0757309 10 g
AAL0757318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0757309 Supplier Thermo Scientific Chemicals Supplier No. L0757309
Only null left

CAS: 29682-39-1 | C6H3BrClNO2 | 236.449 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29682-39-1
Molecular Formula C6H3BrClNO2
Molecular Weight (g/mol) 236.449
MDL Number MFCD00051514
InChI Key YKSXEJZFIQAUHJ-UHFFFAOYSA-N
Synonym 4-bromo-3-chloronitrobenzene, benzene,1-bromo-2-chloro-4-nitro, pubchem22601, acmc-209vlc, 2-chloro-4-nitro-bromobenzene, 4-bromo-3-chloro-nitrobenzene, bromo-2-chloro-4-nitrobenzene, 1-bromanyl-2-chloranyl-4-nitro-benzene
PubChem CID 34667
IUPAC Name 1-bromo-2-chloro-4-nitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Cl)Br

Specifications

Melting Point 59°C to 62°C
Quantity 10 g
UN Number UN2811
Beilstein 2253107
Formula Weight 236.45
Percent Purity 98%
Chemical Name or Material 1-Bromo-2-chloro-4-nitrobenzene
Hazard Category H302+H312+H332-H315-H319-H335-H373
Hazard Statement GHS H Statement
H331-H373-H302-H312-H315-H319-H335
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 249-773-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only