missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2-chloro-3-fluorobenzene 98.0+%, TCI America™
$70.84 - $285.21
Chemical Identifiers
| CAS | 883499-24-9 |
|---|---|
| Molecular Formula | C6H3BrClF |
| Molecular Weight (g/mol) | 209.442 |
| MDL Number | MFCD04038306 |
| InChI Key | GTIWFNAUYPVPAT-UHFFFAOYSA-N |
| PubChem CID | 3739301 |
| IUPAC Name | 1-bromo-2-chloro-3-fluorobenzene |
| SMILES | C1=CC(=C(C(=C1)Br)Cl)F |
Chemical Identifiers
| 883499-24-9 | |
| 209.442 | |
| GTIWFNAUYPVPAT-UHFFFAOYSA-N | |
| 1-bromo-2-chloro-3-fluorobenzene |
| C6H3BrClF | |
| MFCD04038306 | |
| 3739301 | |
| C1=CC(=C(C(=C1)Br)Cl)F |
Specifications
| 883499-24-9 | |
| C6H3BrClF | |
| 5 g | |
| C1=CC(=C(C(=C1)Br)Cl)F | |
| 209.442 | |
| 209.44 | |
| Crystals |
| White-Yellow | |
| MFCD04038306 | |
| GTIWFNAUYPVPAT-UHFFFAOYSA-N | |
| 1-bromo-2-chloro-3-fluorobenzene | |
| 3739301 | |
| ≥98.0% (GC) | |
| 1-Bromo-2-chloro-3-fluorobenzene |
Safety and Handling
TSCA : No