missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-butyne 96.0+%, TCI America™

Catalog No. p-7119540
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21901G 1 g
B21905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21901G Supplier TCI America Supplier No. B21901G
Only null left

Chemical Identifiers

CAS 3355-28-0
Molecular Formula C4H5Br
Molecular Weight (g/mol) 132.988
MDL Number MFCD00190233
InChI Key LNNXOEHOXSYWLD-UHFFFAOYSA-N
Synonym 1-bromo-2-butyne, 2-butyne, 1-bromo, 2-butynyl bromide, 1-bromo-but-2-yne, bromo-2-butyne, 1-bromo2-butyne, but-2-ynylbromide, l-bromo-2-butine, 1-bromo-2-butine, 4-bromo-2-butyne
PubChem CID 2756862
IUPAC Name 1-bromobut-2-yne
SMILES CC#CCBr

Specifications

Color Yellow
Quantity 1 g
UN Number 1993
Formula Weight 132.99
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-butyne