missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-(bromomethyl)naphthalene 97.0+%, TCI America™

Catalog No. B16071G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16071G 1 g
B16075G 5 g
B160725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B16071G Supplier TCI America Supplier No. B16071G

Chemical Identifiers

CAS 37763-43-2
Molecular Formula C11H8Br2
Molecular Weight (g/mol) 299.99
MDL Number MFCD00046369
InChI Key DQTOCXIHYIQHCK-UHFFFAOYSA-N
Synonym 1-bromo-2-bromomethyl naphthalene, naphthalene, 1-bromo-2-bromomethyl, 1-bromo-2-bromomethyl-naphthalene, acmc-209ivc, bromobromomethylnaphtalene, 3-05-00-01633 beilstein handbook reference, wln: l66j be c1e, 1-bromo-2-bromethylnaphthalene, 2-bromomethyl-1-bromonaphthalene
PubChem CID 37828
IUPAC Name 1-bromo-2-(bromomethyl)naphthalene
SMILES BrCC1=CC=C2C=CC=CC2=C1Br

Specifications

Melting Point 108°C
Color White-Yellow
Quantity 1 g
Formula Weight 299.99
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-2-(bromomethyl)naphthalene
RTECSNumber QJ1550000
TSCA No