missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-(bromomethyl)naphthalene, 98%, Thermo Scientific Chemicals

Catalog No. AAH5024003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5024003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5024003 Supplier Thermo Scientific Chemicals Supplier No. H5024003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 37763-43-2 | Molecular Formula: C11H8Br2 | Molecular Weight: 299.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37763-43-2
Molecular Formula C11H8Br2
Molecular Weight (g/mol) 299.99
MDL Number MFCD00046369
InChI Key DQTOCXIHYIQHCK-UHFFFAOYSA-N
Synonym 1-bromo-2-bromomethyl naphthalene, naphthalene, 1-bromo-2-bromomethyl, 1-bromo-2-bromomethyl-naphthalene, acmc-209ivc, bromobromomethylnaphtalene, 3-05-00-01633 beilstein handbook reference, wln: l66j be c1e, 1-bromo-2-bromethylnaphthalene, 2-bromomethyl-1-bromonaphthalene
PubChem CID 37828
IUPAC Name 1-bromo-2-(bromomethyl)naphthalene
SMILES BrCC1=CC=C2C=CC=CC2=C1Br

Specifications

Melting Point 108°C
Quantity 1 g
UN Number UN3261
Formula Weight 299.99
Percent Purity 98%
Chemical Name or Material 1-Bromo-2-(bromomethyl)naphthalene
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
RTECSNumber QJ1550000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only