missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,6-difluorobenzene 98.0+%, TCI America™

Catalog No. D19435G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19435G 5 g
D194325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D19435G Supplier TCI America Supplier No. D19435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64248-56-2
Molecular Formula C6H3BrF2
Molecular Weight (g/mol) 192.991
MDL Number MFCD00009894
InChI Key HRZTZLCMURHWFY-UHFFFAOYSA-N
Synonym 1-bromo-2,6-difluorobenzene, 2,6-difluorobromobenzene, benzene, 2-bromo-1,3-difluoro, 2-bromo-1,3-difluoro-benzene, m-difluorobromobenzene, pubchem3034, acmc-1bc5o, 2,6-difluoro-bromobenzene, 1-bromo-2,6-diflurobenzene
PubChem CID 123557
IUPAC Name 2-bromo-1,3-difluorobenzene
SMILES C1=CC(=C(C(=C1)F)Br)F

Specifications

Color Yellow
Boiling Point 137°C
Quantity 5 g
UN Number 1993
Formula Weight 192.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2,6-difluorobenzene