missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,5-difluoro-4-nitrobenzene, 97%, Thermo Scientific Chemicals

Catalog No. AAA1867006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1867006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1867006 Supplier Thermo Scientific Chemicals Supplier No. A1867006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 167415-27-2 | Molecular Formula: C6H2BrF2NO2 | Molecular Weight: 237.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 167415-27-2
Molecular Formula C6H2BrF2NO2
Molecular Weight (g/mol) 237.99
MDL Number MFCD00042184
InChI Key GJFYMYJYPARISZ-UHFFFAOYSA-N
Synonym 4-bromo-2,5-difluoronitrobenzene, 4-bromo-2,5-difluoro-1-nitrobenzene, benzene, 1-bromo-2,5-difluoro-4-nitro, buttpark 29\01-58, 1-bromo-2,5-difluoro-4-nitro-benzene, pubchem8498, zlchem 1046, acmc-209dwg, intermediates-zcf02256, ksc495e4h
PubChem CID 2736286
IUPAC Name 1-bromo-2,5-difluoro-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC(F)=C(Br)C=C1F

Specifications

Melting Point 55°C to 57°C
Quantity 5 g
Beilstein 7411347
Formula Weight 237.99
Percent Purity 97%
Chemical Name or Material 1-Bromo-2,5-difluoro-4-nitrobenzene
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only