missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,5-dichlorobenzene 98.0+%, TCI America™

$160.30 - $160.30

Chemical Identifiers

CAS 1435-50-3
Molecular Formula C6H3BrCl2
Molecular Weight (g/mol) 225.894
MDL Number MFCD00018505
InChI Key OVXVQBCRONSPDC-UHFFFAOYSA-N
Synonym 1-bromo-2,5-dichlorobenzene, benzene, 2-bromo-1,4-dichloro, 2,5-dichlorobromobenzene, 2-bromo-1,4-dichloro-benzene, benzene, brominated chlorinated, brominated chlorinated benzene, benzene, brominated and chlorinated, pubchem3609, acmc-1bze4, intermediates-zcf02095
PubChem CID 15033
IUPAC Name 2-bromo-1,4-dichlorobenzene
SMILES C1=CC(=C(C=C1Cl)Br)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B098125G
View Documents
TCI America
B098125G
25 g
Each for $160.30
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1435-50-3
225.894
OVXVQBCRONSPDC-UHFFFAOYSA-N
15033
C1=CC(=C(C=C1Cl)Br)Cl
C6H3BrCl2
MFCD00018505
1-bromo-2,5-dichlorobenzene, benzene, 2-bromo-1,4-dichloro, 2,5-dichlorobromobenzene, 2-bromo-1,4-dichloro-benzene, benzene, brominated chlorinated, brominated chlorinated benzene, benzene, brominated and chlorinated, pubchem3609, acmc-1bze4, intermediates-zcf02095
2-bromo-1,4-dichlorobenzene

Specifications

1435-50-3
120°C
MFCD00018505
1-bromo-2,5-dichlorobenzene, benzene, 2-bromo-1,4-dichloro, 2,5-dichlorobromobenzene, 2-bromo-1,4-dichloro-benzene, benzene, brominated chlorinated, brominated chlorinated benzene, benzene, brominated and chlorinated, pubchem3609, acmc-1bze4, intermediates-zcf02095
C1=CC(=C(C=C1Cl)Br)Cl
225.894
225.89
Crystalline Lumps
White-Yellow
C6H3BrCl2
25 g
OVXVQBCRONSPDC-UHFFFAOYSA-N
2-bromo-1,4-dichlorobenzene
15033
≥98.0% (GC)
1-Bromo-2,5-dichlorobenzene
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'