missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,5-dichlorobenzene 98.0+%, TCI America™

Catalog No. B098125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B098125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B098125G Supplier TCI America Supplier No. B098125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1435-50-3
Molecular Formula C6H3BrCl2
Molecular Weight (g/mol) 225.894
MDL Number MFCD00018505
InChI Key OVXVQBCRONSPDC-UHFFFAOYSA-N
Synonym 1-bromo-2,5-dichlorobenzene, benzene, 2-bromo-1,4-dichloro, 2,5-dichlorobromobenzene, 2-bromo-1,4-dichloro-benzene, benzene, brominated chlorinated, brominated chlorinated benzene, benzene, brominated and chlorinated, pubchem3609, acmc-1bze4, intermediates-zcf02095
PubChem CID 15033
IUPAC Name 2-bromo-1,4-dichlorobenzene
SMILES C1=CC(=C(C=C1Cl)Br)Cl

Specifications

Color White-Yellow
Boiling Point 120°C
Quantity 25 g
Formula Weight 225.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Bromo-2,5-dichlorobenzene