missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,5-dichloro-3-fluorobenzene, 97%, Thermo Scientific Chemicals

$436.03 - $436.03

Chemical Identifiers

CAS 202865-57-4
Molecular Formula C6H2BrCl2F
Molecular Weight (g/mol) 243.88
MDL Number MFCD00142585
InChI Key CAYJMDVKWMVOLG-UHFFFAOYSA-N
Synonym 2,5-dichloro-3-fluorobromobenzene, 3-bromo-2,5-dichloro-1-fluorobenzene, pubchem3211, acmc-1cm10, #, 2-bromo-6-fluoro-1,4-phenylene dichloride, benzene, 1-bromo-2,5-dichloro-3-fluoro, 1-bromanyl-2,5-bis chloranyl-3-fluoranyl-benzene
PubChem CID 618513
IUPAC Name 1-bromo-2,5-dichloro-3-fluorobenzene
SMILES FC1=CC(Cl)=CC(Br)=C1Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1671314
View Documents
Thermo Scientific Chemicals
L1671314
25 g
Each for $436.03
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

202865-57-4
243.88
CAYJMDVKWMVOLG-UHFFFAOYSA-N
618513
FC1=CC(Cl)=CC(Br)=C1Cl
C6H2BrCl2F
MFCD00142585
2,5-dichloro-3-fluorobromobenzene, 3-bromo-2,5-dichloro-1-fluorobenzene, pubchem3211, acmc-1cm10, #, 2-bromo-6-fluoro-1,4-phenylene dichloride, benzene, 1-bromo-2,5-dichloro-3-fluoro, 1-bromanyl-2,5-bis chloranyl-3-fluoranyl-benzene
1-bromo-2,5-dichloro-3-fluorobenzene

Specifications

202865-57-4
1.5757
25 g
CAYJMDVKWMVOLG-UHFFFAOYSA-N
1-bromo-2,5-dichloro-3-fluorobenzene
618513
97%
C6H2BrCl2F
MFCD00142585
2,5-dichloro-3-fluorobromobenzene, 3-bromo-2,5-dichloro-1-fluorobenzene, pubchem3211, acmc-1cm10, #, 2-bromo-6-fluoro-1,4-phenylene dichloride, benzene, 1-bromo-2,5-dichloro-3-fluoro, 1-bromanyl-2,5-bis chloranyl-3-fluoranyl-benzene
FC1=CC(Cl)=CC(Br)=C1Cl
243.88
243.89
1-Bromo-2,5-dichloro-3-fluorobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only