missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,5-dichloro-3-fluorobenzene, 97%, Thermo Scientific Chemicals

Catalog No. AAL1671314
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1671314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1671314 Supplier Thermo Scientific Chemicals Supplier No. L1671314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 202865-57-4 | Molecular Formula: C6H2BrCl2F | Molecular Weight: 243.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 202865-57-4
Molecular Formula C6H2BrCl2F
Molecular Weight (g/mol) 243.88
MDL Number MFCD00142585
InChI Key CAYJMDVKWMVOLG-UHFFFAOYSA-N
Synonym 2,5-dichloro-3-fluorobromobenzene, 3-bromo-2,5-dichloro-1-fluorobenzene, pubchem3211, acmc-1cm10, #, 2-bromo-6-fluoro-1,4-phenylene dichloride, benzene, 1-bromo-2,5-dichloro-3-fluoro, 1-bromanyl-2,5-bis chloranyl-3-fluoranyl-benzene
PubChem CID 618513
IUPAC Name 1-bromo-2,5-dichloro-3-fluorobenzene
SMILES FC1=CC(Cl)=CC(Br)=C1Cl

Specifications

Refractive Index 1.5757
Quantity 25 g
Formula Weight 243.89
Percent Purity 97%
Chemical Name or Material 1-Bromo-2,5-dichloro-3-fluorobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only