missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,4-difluorobenzene 98.0+%, TCI America™

Catalog No. D190925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D190925G 25 g
D1909500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D190925G Supplier TCI America Supplier No. D190925G

Chemical Identifiers

CAS 348-57-2
Molecular Formula C6H3BrF2
Molecular Weight (g/mol) 192.99
MDL Number MFCD00000330
InChI Key MGHBDQZXPCTTIH-UHFFFAOYSA-N
Synonym 2,4-difluorobromobenzene, benzene, 1-bromo-2,4-difluoro, 1-bromo-2,4-difluoro-benzene, bromo-2,4-difluorobenzene, 2,4-difluoro-1-bromobenzene, attercop-chm at130629, fr cf de, pubchem2301, 2.4-difluorobromobenzene, acmc-1adm3
PubChem CID 67674
IUPAC Name 1-bromo-2,4-difluorobenzene
SMILES FC1=CC=C(Br)C(F)=C1

Specifications

Color Yellow
Boiling Point 146°C
Quantity 25 g
UN Number 1993
Formula Weight 192.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2,4-difluorobenzene