missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,4-difluorobenzene, 98+%

Catalog No. AAA1409836
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1409836 500 g
AAA1409814 25 g
AAA1409822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1409836 Supplier Thermo Scientific Chemicals Supplier No. A1409836
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 348-57-2 | C6H3BrF2 | 192.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 348-57-2
Molecular Formula C6H3BrF2
Molecular Weight (g/mol) 192.99
MDL Number MFCD00000330
InChI Key MGHBDQZXPCTTIH-UHFFFAOYSA-N
Synonym 2,4-difluorobromobenzene, benzene, 1-bromo-2,4-difluoro, 1-bromo-2,4-difluoro-benzene, bromo-2,4-difluorobenzene, 2,4-difluoro-1-bromobenzene, attercop-chm at130629, fr cf de, pubchem2301, 2.4-difluorobromobenzene, acmc-1adm3
PubChem CID 67674
IUPAC Name 1-bromo-2,4-difluorobenzene
SMILES FC1=CC=C(Br)C(F)=C1

Specifications

Melting Point -4°C
Density 1.708
Boiling Point 145°C to 146°C
Flash Point 51°C (123°F)
Refractive Index 1.505
Quantity 500 g
UN Number UN1993
Beilstein 1680892
Formula Weight 192.99
Percent Purity ≥98%
Chemical Name or Material 1-Bromo-2,4-difluorobenzene
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 206-479-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only