missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,4-difluorobenzene, 98+%

Catalog No. p-73803
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC249360100 10 mL Glass bottle
AC249360500 50 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC249360100 Supplier Thermo Scientific Chemicals Supplier No. 249360100
Only null left

CAS: 348-57-2 | C6H3BrF2 | 192.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 348-57-2
Molecular Formula C6H3BrF2
Molecular Weight (g/mol) 192.99
MDL Number MFCD00000330
InChI Key MGHBDQZXPCTTIH-UHFFFAOYSA-N
Synonym 2,4-difluorobromobenzene, benzene, 1-bromo-2,4-difluoro, 1-bromo-2,4-difluoro-benzene, bromo-2,4-difluorobenzene, 2,4-difluoro-1-bromobenzene, attercop-chm at130629, fr cf de, pubchem2301, 2.4-difluorobromobenzene, acmc-1adm3
PubChem CID 67674
IUPAC Name 1-bromo-2,4-difluorobenzene
SMILES FC1=CC=C(Br)C(F)=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -4°C
Color Colorless to Brown
Density 1.7080g/mL
Boiling Point 145°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5040 to 1.506
Linear Formula BrC6H3F2
Quantity 10 mL
Specific Gravity 1.708
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in most common organic solvents
Formula Weight 192.99
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 1-Bromo-2, 4-difluorobenzene, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-479-4
TSCA TSCA

RUO – Research Use Only