missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,4-dichlorobenzene 98.0+%, TCI America™

Catalog No. p-7119537
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21875G 5 g
B218725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21875G Supplier TCI America Supplier No. B21875G

Chemical Identifiers

CAS 1193-72-2
Molecular Formula C6H3BrCl2
Molecular Weight (g/mol) 225.894
MDL Number MFCD00018333
InChI Key ISHYFWKKWKXXPL-UHFFFAOYSA-N
Synonym 2,4-dichlorobromobenzene, bromo 2--4-dichlorobenzene, 1-bromo-2,4-dichloro-benzene, benzene, 1-bromo-2,4-dichloro, pubchem3608, 2,4-dichlorobromobenze, 2,4-dichloro-bromobenzene, acmc-209a2k, 2,4-dichloro-1-bromobenzene, ksc490m9l
PubChem CID 70947
IUPAC Name 1-bromo-2,4-dichlorobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)Br

Specifications

Color White-Yellow
Boiling Point 235°C
Quantity 5 g
Formula Weight 225.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Bromo-2,4-dichlorobenzene