missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,4-dichlorobenzene, 98%

Catalog No. AAA1229822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1229822 100 g
AAA1229806 5 g
AAA1229814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1229822 Supplier Thermo Scientific Chemicals Supplier No. A1229822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1193-72-2 | C6H3BrCl2 | 225.894 g/mol

1-Bromo-2,4-dichlorobenzene is used to produce 2,4,4'-trichloro-biphenyl. This reaction will need aq. Na2CO3 and solvents dioxane, ethanol. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2,4-dichlorobenzene is used to produce 2,4,4′-trichloro-biphenyl. This reaction will need aq. Na2CO3 and solvents dioxane, ethanol. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1193-72-2
Molecular Formula C6H3BrCl2
Molecular Weight (g/mol) 225.894
MDL Number MFCD00018333
InChI Key ISHYFWKKWKXXPL-UHFFFAOYSA-N
Synonym 2,4-dichlorobromobenzene, bromo 2--4-dichlorobenzene, 1-bromo-2,4-dichloro-benzene, benzene, 1-bromo-2,4-dichloro, pubchem3608, 2,4-dichlorobromobenze, 2,4-dichloro-bromobenzene, acmc-209a2k, 2,4-dichloro-1-bromobenzene, ksc490m9l
PubChem CID 70947
IUPAC Name 1-bromo-2,4-dichlorobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)Br

Specifications

Melting Point 26°C to 27°C
Boiling Point 235°C
Quantity 100 g
Beilstein 1635733
Solubility Information Slightly soluble in water.
Formula Weight 225.91
Percent Purity 98%
Chemical Name or Material 1-Bromo-2,4-dichlorobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-778-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only