missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2,4,5-trimethoxybenzene 98.0+%, TCI America™
$209.84 - $635.31
Chemical Identifiers
| CAS | 20129-11-7 |
|---|---|
| Molecular Formula | C9H11BrO3 |
| Molecular Weight (g/mol) | 247.088 |
| MDL Number | MFCD00094684 |
| InChI Key | SFEPXIIFUHNCDO-UHFFFAOYSA-N |
| PubChem CID | 285896 |
| IUPAC Name | 1-bromo-2,4,5-trimethoxybenzene |
| SMILES | COC1=CC(=C(C=C1OC)Br)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B4721200MG
|
TCI America
B4721200MG |
200 mg |
Each for $209.84
|
|
|||||
|
B47211G
|
TCI America
B47211G |
1 g |
Each for $635.31
|
|
|||||
Chemical Identifiers
| 20129-11-7 | |
| 247.088 | |
| SFEPXIIFUHNCDO-UHFFFAOYSA-N | |
| 1-bromo-2,4,5-trimethoxybenzene |
| C9H11BrO3 | |
| MFCD00094684 | |
| 285896 | |
| COC1=CC(=C(C=C1OC)Br)OC |
Specifications
| 20129-11-7 | |
| White-Yellow | |
| C9H11BrO3 | |
| 200 mg | |
| COC1=CC(=C(C=C1OC)Br)OC | |
| 247.088 | |
| 247.09 | |
| Crystalline Powder |
| 55°C | |
| 263°C | |
| MFCD00094684 | |
| SFEPXIIFUHNCDO-UHFFFAOYSA-N | |
| 1-bromo-2,4,5-trimethoxybenzene | |
| 285896 | |
| ≥98.0% (GC) | |
| 1-Bromo-2,4,5-trimethoxybenzene |
Safety and Handling
TSCA : No