missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,4,5-trimethoxybenzene 98.0+%, TCI America™

Catalog No. B4721200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4721200MG 200 mg
B47211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4721200MG Supplier TCI America Supplier No. B4721200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20129-11-7
Molecular Formula C9H11BrO3
Molecular Weight (g/mol) 247.088
MDL Number MFCD00094684
InChI Key SFEPXIIFUHNCDO-UHFFFAOYSA-N
PubChem CID 285896
IUPAC Name 1-bromo-2,4,5-trimethoxybenzene
SMILES COC1=CC(=C(C=C1OC)Br)OC

Specifications

Melting Point 55°C
Color White-Yellow
Boiling Point 263°C
Quantity 200 mg
Formula Weight 247.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-2,4,5-trimethoxybenzene