missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,3-dichlorobenzene 99.0+%, TCI America™

Catalog No. p-7118777
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B09805G 5 g
B098025G 25 g
2 options

Catalog No. B09805G

Supplier: TCI America B09805G

Only null left

Chemical Identifiers

CAS 56961-77-4
Molecular Formula C6H3BrCl2
Molecular Weight (g/mol) 225.89
MDL Number MFCD00000536
InChI Key HVKCZUVMQPUWSX-UHFFFAOYSA-N
Synonym 2,3-dichlorobromobenzene, benzene, 1-bromo-2,3-dichloro, benzene, bromodichloro, 2,3-dichloro-1-bromobenzene, 1-bromo-2,3-dichloro-benzene, attercop-chm at113305, bromodichlorobenzene, pubchem3607, monobromodichlorobenzene, acmc-209lv2
PubChem CID 42066
IUPAC Name 1-bromo-2,3-dichlorobenzene
SMILES ClC1=CC=CC(Br)=C1Cl

Specifications

Melting Point 60°C
Boiling Point 243°C
Quantity 5 g
Formula Weight 225.89
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-2,3-dichlorobenzene